Product Details

SNP ID
rs121434508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85654139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTCATAGCAAACAGTCTTTTT[C/T]GACGCATCTTGAATGTGACAAAGGC
Phenotype
MIM: 606444 MIM: 612988
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CREBZF PubMed Links

Gene Details

Gene
CREBZF
Gene Name
CREB/ATF bZIP transcription factor
There are no transcripts associated with this gene.

Gene
TMEM126A
Gene Name
transmembrane protein 126A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244735.1 248 UTR 5 NP_001231664.1
NM_032273.3 248 Intron NP_115649.1

View Full Product Details