Product Details

SNP ID
rs138108997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4545425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGACCTGGTTCTGTCTACTTCC[A/C]CTATACCCAAACTTCTGGGAATCTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52M1 PubMed Links

Gene Details

Gene
OR52M1
Gene Name
olfactory receptor family 52 subfamily M member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004137.1 235 Missense Mutation ACT,CCT T79P NP_001004137.1

View Full Product Details