Product Details

SNP ID
rs138708788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:7089933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAGGTCGCGACCGTGACTACGG[A/G]GATCATCTGAGCAGAGGCTCCCATC
Phenotype
MIM: 609665 MIM: 605444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRP14 PubMed Links

Gene Details

Gene
NLRP14
Gene Name
NLR family pyrin domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176822.3 1000 Intron NP_789792.1
XM_011520044.1 1000 Intron XP_011518346.1
Gene
RBMXL2
Gene Name
RNA binding motif protein, X-linked like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014469.4 1000 Silent Mutation GGA,GGG G271G NP_055284.3

View Full Product Details