Product Details

SNP ID
rs139090970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66473371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCCGCATCCTCTGTGACCGCC[A/G]GCCACCCTATACTGCCCGCATCTAT
Phenotype
MIM: 606818 MIM: 609827
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPP3 PubMed Links

Gene Details

Gene
DPP3
Gene Name
dipeptidyl peptidase 3
There are no transcripts associated with this gene.

Gene
LOC101928069
Gene Name
uncharacterized LOC101928069
There are no transcripts associated with this gene.

Gene
PELI3
Gene Name
pellino E3 ubiquitin protein ligase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098510.1 518 Missense Mutation CAG,CGG Q172R NP_001091980.1
NM_001243135.1 518 Missense Mutation CAG,CGG Q133R NP_001230064.1
NM_001243136.1 518 Missense Mutation CAG,CGG Q89R NP_001230065.1
NM_145065.2 518 Missense Mutation CAG,CGG Q196R NP_659502.2
XM_011544884.2 518 Missense Mutation CAG,CGG Q196R XP_011543186.1
XM_017017465.1 518 Missense Mutation CAG,CGG Q47R XP_016872954.1

View Full Product Details