Product Details

SNP ID
rs139297420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117286204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGACATATTTCCTGTCCTGGGAA[A/G]AATGGAGACCAAAGTGATATTAAGA
Phenotype
MIM: 604252 MIM: 614263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BACE1 PubMed Links

Gene Details

Gene
BACE1
Gene Name
beta-secretase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207048.1 5254 UTR 3 NP_001193977.1
NM_001207049.1 5254 UTR 3 NP_001193978.1
NM_012104.4 5254 UTR 3 NP_036236.1
NM_138971.3 5254 UTR 3 NP_620427.1
NM_138972.3 5254 UTR 3 NP_620428.1
NM_138973.3 5254 UTR 3 NP_620429.1
Gene
BACE1-AS
Gene Name
BACE1 antisense RNA
There are no transcripts associated with this gene.

Gene
RNF214
Gene Name
ring finger protein 214
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077239.1 5254 Intron NP_001070707.1
NM_001278249.1 5254 Intron NP_001265178.1
NM_207343.3 5254 Intron NP_997226.2

View Full Product Details