Product Details

SNP ID
rs141171319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6716869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGGGGGTTGCCATCCCAGAGGT[A/G]AGCAAAGTAGTAGACGTGAGTATTG
Phenotype
MIM: 616121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GVINP1 PubMed Links

Gene Details

Gene
GVINP1
Gene Name
GTPase, very large interferon inducible pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details