Product Details

SNP ID
rs141396989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:110579439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTAGGCCCTGAGTCTGGGCTGGT[C/T]GCATCCTCTAGAGTCCAAGAGCTGG
Phenotype
MIM: 609568
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP20 PubMed Links

Gene Details

Gene
ARHGAP20
Gene Name
Rho GTPase activating protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258415.1 3613 Silent Mutation GCA,GCG A1146A NP_001245344.1
NM_001258416.1 3613 Silent Mutation GCA,GCG A1143A NP_001245345.1
NM_001258417.1 3613 Silent Mutation GCA,GCG A1133A NP_001245346.1
NM_001258418.1 3613 Silent Mutation GCA,GCG A1133A NP_001245347.1
NM_020809.3 3613 Silent Mutation GCA,GCG A1169A NP_065860.2
XM_005271628.3 3613 Silent Mutation GCA,GCG A712A XP_005271685.1
XM_006718893.3 3613 Silent Mutation GCA,GCG A1169A XP_006718956.1

View Full Product Details