Product Details

SNP ID
rs141802468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68713367 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATTGCCCAATTGGCCCTTCCCA[A/G]TTTTTGGCTGGAAAGCTTCTGGATT
Phenotype
MIM: 604374
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TESMIN PubMed Links

Gene Details

Gene
TESMIN
Gene Name
testis expressed metallothionein like protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039656.1 1047 Intron NP_001034745.1
NM_004923.3 1047 Missense Mutation ACT,ATT T354I NP_004914.2
XM_011545402.1 1047 Missense Mutation ACT,ATT T354I XP_011543704.1
XM_011545403.2 1047 Missense Mutation ACT,ATT T354I XP_011543705.1
XM_017018588.1 1047 Missense Mutation ACT,ATT T288I XP_016874077.1

View Full Product Details