Product Details

SNP ID
rs141895562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114063789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCTTCATCTCGAAGCATTCCAG[C/T]GAGGAGAGTGGGTATGCCAGTGTGG
Phenotype
MIM: 176797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZBTB16 PubMed Links

Gene Details

Gene
ZBTB16
Gene Name
zinc finger and BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018011.1 1193 Silent Mutation AGC,AGT S163S NP_001018011.1
NM_006006.4 1193 Silent Mutation AGC,AGT S163S NP_005997.2
XM_005271658.4 1193 Silent Mutation AGC,AGT S163S XP_005271715.1
XM_017018257.1 1193 Silent Mutation AGC,AGT S163S XP_016873746.1
XM_017018258.1 1193 Silent Mutation AGC,AGT S163S XP_016873747.1
XM_017018259.1 1193 Silent Mutation AGC,AGT S163S XP_016873748.1

View Full Product Details