Product Details

SNP ID
rs142115592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114063357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGAACCCTAGCCACCCCACGGG[A/G]CTACTGTGCAAGGCCAACCAGATGC
Phenotype
MIM: 176797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB16 PubMed Links

Gene Details

Gene
ZBTB16
Gene Name
zinc finger and BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018011.1 761 Silent Mutation GGA,GGG G19G NP_001018011.1
NM_006006.4 761 Silent Mutation GGA,GGG G19G NP_005997.2
XM_005271658.4 761 Silent Mutation GGA,GGG G19G XP_005271715.1
XM_017018257.1 761 Silent Mutation GGA,GGG G19G XP_016873746.1
XM_017018258.1 761 Silent Mutation GGA,GGG G19G XP_016873747.1
XM_017018259.1 761 Silent Mutation GGA,GGG G19G XP_016873748.1

View Full Product Details