Product Details

SNP ID
rs142278050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17091341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGATTTTAACTTACCATTTCA[C/T]TGAATGTCGGATTCCTCGTTTTTCG
Phenotype
MIM: 603601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIK3C2A PubMed Links

Gene Details

Gene
PIK3C2A
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321378.1 5101 Missense Mutation AAT,AGT N1624S NP_001308307.1
NM_001321380.1 5101 Missense Mutation AAT,AGT N1244S NP_001308309.1
NM_002645.3 5101 Missense Mutation AAT,AGT N1624S NP_002636.2
XM_011520186.2 5101 Missense Mutation AAT,AGT N1568S XP_011518488.1
XM_017017924.1 5101 Missense Mutation AAT,AGT N1624S XP_016873413.1

View Full Product Details