Product Details

SNP ID
rs142746293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55796026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCATATCTCTGGGGCATGTTTGG[C/T]CCCTTGGTACTCCTTTGTTATGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5D14 PubMed Links

Gene Details

Gene
OR5D14
Gene Name
olfactory receptor family 5 subfamily D member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004735.1 471 Silent Mutation GGC,GGT G157G NP_001004735.1

View Full Product Details