Product Details

SNP ID
rs142883872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82914746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCACGGTGTGAACACATGAACA[A/G]AAGACGAAAATTTCTTCTAGCCTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDIAS PubMed Links

Gene Details

Gene
DDIAS
Gene Name
DNA damage induced apoptosis suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145018.3 114 Missense Mutation AAA,AGA K3R NP_659455.3
XM_011544835.1 114 Missense Mutation AAA,AGA K3R XP_011543137.1
XM_011544836.2 114 Missense Mutation AAA,AGA K3R XP_011543138.1
XM_011544837.1 114 Intron XP_011543139.1
XM_011544838.2 114 UTR 5 XP_011543140.1
XM_017017352.1 114 UTR 5 XP_016872841.1
XM_017017353.1 114 UTR 5 XP_016872842.1

View Full Product Details