Product Details
- SNP ID
-
rs143254060
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:6319392 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAAAGGGCCCTTCGGAGGCTCTGT[A/G]CCTTCTGCAATCCGGTGCGCCGCAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRKCDBP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1051992] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRKCDBP
- Gene Name
- protein kinase C delta binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145040.2 |
591 |
Missense Mutation |
GCA,GTA |
A186V |
NP_659477.2 |
View Full Product Details