Product Details
- SNP ID
-
rs143630318
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118307293 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCTTATTTATTTTCTAGTTGGC[A/G]TTTGGGGGCAAGATGGTGAGATATG
- Phenotype
-
MIM: 186830
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD3E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1126924] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD3E
- Gene Name
- CD3e molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000733.3 |
311 |
Missense Mutation |
ATT,GTT |
I19V |
NP_000724.1 |
View Full Product Details