Product Details

SNP ID
rs144522181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6319337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCGAGGCGGCTTGACCGGGGT[A/G]GGCGGTGGCGCTGCAGGGCCTTTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKCDBP PubMed Links

Gene Details

Gene
PRKCDBP
Gene Name
protein kinase C delta binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145040.2 646 Silent Mutation CCC,CCT P204P NP_659477.2

View Full Product Details