Product Details
- SNP ID
-
rs144571267
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:55796132 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCTCATCTCTGTGTCTGGCTCT[A/G]ATATACTCATCCCCCACCTGCTGCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR5D14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs68042892] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR5D14
- Gene Name
- olfactory receptor family 5 subfamily D member 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004735.1 |
577 |
Missense Mutation |
AAT,GAT |
N193D |
NP_001004735.1 |
View Full Product Details