Product Details

SNP ID
rs145143636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65266634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGAGGAGGGAATGGTAGGCGA[A/G]CTTATAGCCTTCTGCACCAGCACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLA2 PubMed Links

Gene Details

Gene
POLA2
Gene Name
DNA polymerase alpha 2, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002689.3 512 Silent Mutation GAA,GAG E44E NP_002680.2
XM_011544877.2 512 Silent Mutation GAA,GAG E44E XP_011543179.1
XM_011544878.2 512 Silent Mutation GAA,GAG E44E XP_011543180.1
XM_011544879.1 512 Silent Mutation GAA,GAG E44E XP_011543181.1
XM_011544881.2 512 Silent Mutation GAA,GAG E44E XP_011543183.1
XM_017017452.1 512 Silent Mutation GAA,GAG E44E XP_016872941.1

View Full Product Details