Product Details

SNP ID
rs145297189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123883215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGTGTGCACGGACAATGCTAC[A/G]AGGCATTGCAGTGCATTCTGGTAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM225 PubMed Links

Gene Details

Gene
TMEM225
Gene Name
transmembrane protein 225
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013743.2 940 Missense Mutation CGT,TGT R201C NP_001013765.2
XM_006718832.3 940 Missense Mutation CGT,TGT R151C XP_006718895.1
XM_011542802.2 940 Missense Mutation CGT,TGT R156C XP_011541104.1
XM_017017653.1 940 Intron XP_016873142.1

View Full Product Details