Product Details

SNP ID
rs145563235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119335971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGGCTCTCTCGCCTAGCACTGGG[C/G]TCAGAGGCCTGGCGCCGAGTCTGGA
Phenotype
MIM: 608752 MIM: 606227 MIM: 606130
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1QTNF5 PubMed Links

Gene Details

Gene
C1QTNF5
Gene Name
C1q and tumor necrosis factor related protein 5
There are no transcripts associated with this gene.

Gene
MFRP
Gene Name
membrane frizzled-related protein
There are no transcripts associated with this gene.

Gene
RNF26
Gene Name
ring finger protein 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032015.4 1472 Silent Mutation GGC,GGG G283G NP_114404.1

View Full Product Details