Product Details

SNP ID
rs145754283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66294565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTACATCCCGCTGGCCTTCGTC[G/T]CCCTCCTCTACCTCCTCTACCTGGC
Phenotype
MIM: 611484
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM151A PubMed Links

Gene Details

Gene
TMEM151A
Gene Name
transmembrane protein 151A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153266.3 431 Missense Mutation GCC,TCC A107S NP_694998.1
Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details