Product Details

SNP ID
rs145832341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:85685362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGGCCTGCAGCTCCTGGCTCA[A/G]GCTGCTGTGTGTCTCCTCTGCCTTA
Phenotype
MIM: 612880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC89 PubMed Links

Gene Details

Gene
CCDC89
Gene Name
coiled-coil domain containing 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152723.2 916 Silent Mutation CTG,TTG L257L NP_689936.1
Gene
SYTL2
Gene Name
synaptotagmin like 2
There are no transcripts associated with this gene.

View Full Product Details