Product Details

SNP ID
rs146925616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123939896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCAGGCAACCCTACCATTTCC[G/T]TTGGTGGATGCCTGACTCAACTCTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4D5 PubMed Links

Gene Details

Gene
OR4D5
Gene Name
olfactory receptor family 4 subfamily D member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001965.1 280 Missense Mutation GTT,TTT V94F NP_001001965.1
Gene
OR6T1
Gene Name
olfactory receptor family 6 subfamily T member 1
There are no transcripts associated with this gene.

View Full Product Details