Product Details

SNP ID
rs148055755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134249260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGTCTGGGTAAGCCTCTTTCAC[A/G]ATCTGAAACCAAAACAAAAGCTTTG
Phenotype
MIM: 604773 MIM: 613739 MIM: 610027
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAD8 PubMed Links

Gene Details

Gene
ACAD8
Gene Name
acyl-CoA dehydrogenase family member 8
There are no transcripts associated with this gene.

Gene
THYN1
Gene Name
thymocyte nuclear protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037304.1 489 Silent Mutation ATC,ATT I129I NP_001032381.1
NM_001037305.1 489 Silent Mutation ATC,ATT I129I NP_001032382.1
NM_014174.2 489 Silent Mutation ATC,ATT I129I NP_054893.1
NM_199297.1 489 Silent Mutation ATC,ATT I129I NP_954994.1
NM_199298.1 489 Silent Mutation ATC,ATT I129I NP_954995.1
XM_005271527.3 489 Silent Mutation ATC,ATT I170I XP_005271584.1
XM_005271528.3 489 Silent Mutation ATC,ATT I170I XP_005271585.1
Gene
VPS26B
Gene Name
VPS26, retromer complex component B
There are no transcripts associated with this gene.

View Full Product Details