Product Details

SNP ID
rs148291205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47274764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCACTGACAAGGACACTGGAGT[C/T]ACGCGATATGGCATCTGTGTTAACT
Phenotype
MIM: 603584 MIM: 602423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928943 PubMed Links

Gene Details

Gene
LOC101928943
Gene Name
uncharacterized LOC101928943
There are no transcripts associated with this gene.

Gene
MADD
Gene Name
MAP kinase activating death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135943.1 479 Silent Mutation GTC,GTT V88V NP_001129415.1
NM_001135944.1 479 Silent Mutation GTC,GTT V88V NP_001129416.1
NM_003682.3 479 Silent Mutation GTC,GTT V88V NP_003673.3
NM_130470.2 479 Silent Mutation GTC,GTT V88V NP_569826.2
NM_130471.2 479 Silent Mutation GTC,GTT V88V NP_569827.2
NM_130472.2 479 Silent Mutation GTC,GTT V88V NP_569828.2
NM_130473.2 479 Silent Mutation GTC,GTT V88V NP_569829.2
NM_130474.2 479 Silent Mutation GTC,GTT V88V NP_569830.2
NM_130475.2 479 Silent Mutation GTC,GTT V88V NP_569831.1
NM_130476.2 479 Silent Mutation GTC,GTT V88V NP_569832.2
XM_005253189.2 479 Silent Mutation GTC,GTT V88V XP_005253246.1
XM_005253196.2 479 Silent Mutation GTC,GTT V88V XP_005253253.1
XM_005253199.2 479 Silent Mutation GTC,GTT V88V XP_005253256.1
XM_005253200.2 479 Silent Mutation GTC,GTT V88V XP_005253257.1
XM_005253201.2 479 Silent Mutation GTC,GTT V88V XP_005253258.1
XM_005253203.2 479 Silent Mutation GTC,GTT V88V XP_005253260.1
XM_005253204.2 479 Silent Mutation GTC,GTT V88V XP_005253261.1
XM_005253205.1 479 Silent Mutation GTC,GTT V88V XP_005253262.1
XM_017018478.1 479 Silent Mutation GTC,GTT V88V XP_016873967.1
XM_017018479.1 479 Silent Mutation GTC,GTT V88V XP_016873968.1
XM_017018480.1 479 Silent Mutation GTC,GTT V88V XP_016873969.1
XM_017018481.1 479 Silent Mutation GTC,GTT V88V XP_016873970.1
XM_017018482.1 479 Silent Mutation GTC,GTT V88V XP_016873971.1
XM_017018483.1 479 Silent Mutation GTC,GTT V88V XP_016873972.1
XM_017018484.1 479 Silent Mutation GTC,GTT V88V XP_016873973.1
XM_017018485.1 479 Silent Mutation GTC,GTT V88V XP_016873974.1
XM_017018486.1 479 Silent Mutation GTC,GTT V88V XP_016873975.1
XM_017018487.1 479 Silent Mutation GTC,GTT V88V XP_016873976.1
XM_017018488.1 479 Silent Mutation GTC,GTT V88V XP_016873977.1
XM_017018489.1 479 Silent Mutation GTC,GTT V88V XP_016873978.1
XM_017018490.1 479 Silent Mutation GTC,GTT V88V XP_016873979.1
XM_017018491.1 479 Silent Mutation GTC,GTT V88V XP_016873980.1
XM_017018492.1 479 Silent Mutation GTC,GTT V88V XP_016873981.1
XM_017018493.1 479 Silent Mutation GTC,GTT V88V XP_016873982.1
XM_017018494.1 479 Silent Mutation GTC,GTT V88V XP_016873983.1
XM_017018495.1 479 Silent Mutation GTC,GTT V88V XP_016873984.1
XM_017018496.1 479 Silent Mutation GTC,GTT V88V XP_016873985.1
XM_017018497.1 479 Silent Mutation GTC,GTT V88V XP_016873986.1
XM_017018498.1 479 Silent Mutation GTC,GTT V88V XP_016873987.1
XM_017018499.1 479 Silent Mutation GTC,GTT V88V XP_016873988.1
XM_017018500.1 479 Silent Mutation GTC,GTT V88V XP_016873989.1
XM_017018501.1 479 Silent Mutation GTC,GTT V88V XP_016873990.1
XM_017018502.1 479 Silent Mutation GTC,GTT V88V XP_016873991.1
XM_017018503.1 479 Silent Mutation GTC,GTT V88V XP_016873992.1
XM_017018504.1 479 Silent Mutation GTC,GTT V88V XP_016873993.1
XM_017018505.1 479 Silent Mutation GTC,GTT V88V XP_016873994.1
XM_017018506.1 479 Silent Mutation GTC,GTT V88V XP_016873995.1
XM_017018507.1 479 Silent Mutation GTC,GTT V88V XP_016873996.1
XM_017018508.1 479 Silent Mutation GTC,GTT V88V XP_016873997.1
XM_017018509.1 479 Silent Mutation GTC,GTT V88V XP_016873998.1
XM_017018510.1 479 Silent Mutation GTC,GTT V88V XP_016873999.1
Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
There are no transcripts associated with this gene.

View Full Product Details