Product Details

SNP ID
rs148394048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108672543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATATTTGTATTTTCAGCATTGGA[A/G]GGCACAGAGTAAACACTGCCTGGGC
Phenotype
MIM: 601235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX10 PubMed Links

Gene Details

Gene
DDX10
Gene Name
DEAD-box helicase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004398.3 Intron NP_004389.2

View Full Product Details