Product Details

SNP ID
hCV161526589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14879207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAACACTTCTGGGTCTCTCCAGT[A/T]CTTTTCATCAAAGTGTACAGAATAA
Phenotype
MIM: 608713 MIM: 602047
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP2R1 PubMed Links

Gene Details

Gene
CYP2R1
Gene Name
cytochrome P450 family 2 subfamily R member 1
There are no transcripts associated with this gene.

Gene
PDE3B
Gene Name
phosphodiesterase 3B
There are no transcripts associated with this gene.

View Full Product Details