Product Details

SNP ID
rs149212229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73270472 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGCACTGTGCTGGGAGGCTCA[C/T]CCCTGAACCTACCTTTGTGGACATG
Phenotype
MIM: 602451
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RY6 PubMed Links

Gene Details

Gene
P2RY6
Gene Name
pyrimidinergic receptor P2Y6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277204.1 Intron NP_001264133.1
NM_001277205.1 Intron NP_001264134.1
NM_001277206.1 Intron NP_001264135.1
NM_001277207.1 Intron NP_001264136.1
NM_001277208.1 Intron NP_001264137.1
NM_176796.2 Intron NP_789766.1
NM_176797.2 Intron NP_789767.1
NM_176798.2 Intron NP_789768.1
XM_005274022.3 Intron XP_005274079.1
XM_006718571.3 Intron XP_006718634.1
XM_011545076.2 Intron XP_011543378.1
XM_011545077.2 Intron XP_011543379.1
XM_011545079.2 Intron XP_011543381.1

View Full Product Details