Product Details

SNP ID
rs149371777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55665170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTCATATCAACACCTGAGAAATG[A/G]AAAATCAAAACAATGTGACTGAATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4C6 PubMed Links

Gene Details

Gene
OR4C6
Gene Name
olfactory receptor family 4 subfamily C member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004704.1 4 Missense Mutation AAA,GAA K2E NP_001004704.1

View Full Product Details