Product Details

SNP ID
rs149427006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5046927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCGTCTGTGCTCCACTACATTA[C/T]GCAACCATCTTGACATCCCAAGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52J3 PubMed Links

Gene Details

Gene
OR52J3
Gene Name
olfactory receptor family 52 subfamily J member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001916.2 402 Silent Mutation TAC,TAT Y134Y NP_001001916.2

View Full Product Details