Product Details

SNP ID
rs149851583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55811830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCTGGCCGTGATGGCCTATGAC[C/T]GCTTTGTGGCCATCTGTAACCCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5D18 PubMed Links

Gene Details

Gene
OR5D18
Gene Name
olfactory receptor family 5 subfamily D member 18
There are no transcripts associated with this gene.

Gene
OR5L1
Gene Name
olfactory receptor family 5 subfamily L member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004738.1 364 Missense Mutation CGC,TGC R122C NP_001004738.1

View Full Product Details