Product Details

SNP ID
rs149986878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117288248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATACTGAAGGCAGTAATGTTA[A/G]AGCCATAAAGCACTGTATTTCTCAT
Phenotype
MIM: 604252 MIM: 614263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BACE1 PubMed Links

Gene Details

Gene
BACE1
Gene Name
beta-secretase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207048.1 3210 UTR 3 NP_001193977.1
NM_001207049.1 3210 UTR 3 NP_001193978.1
NM_012104.4 3210 UTR 3 NP_036236.1
NM_138971.3 3210 UTR 3 NP_620427.1
NM_138972.3 3210 UTR 3 NP_620428.1
NM_138973.3 3210 UTR 3 NP_620429.1
Gene
BACE1-AS
Gene Name
BACE1 antisense RNA
There are no transcripts associated with this gene.

Gene
RNF214
Gene Name
ring finger protein 214
There are no transcripts associated with this gene.

View Full Product Details