Product Details

SNP ID
rs150222887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55796055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTACTCCTTTGTTATGCTCTCC[A/G]GTTAAACTTCTCTGGACCTAATGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5D14 PubMed Links

Gene Details

Gene
OR5D14
Gene Name
olfactory receptor family 5 subfamily D member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004735.1 500 Missense Mutation CAG,CGG Q167R NP_001004735.1

View Full Product Details