Product Details

SNP ID
rs150312046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118137107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTGTCATTCCCCGAGGAGCTCA[C/T]GAGACACTCCTTCCTGGAGAGGGAG
Phenotype
MIM: 608256
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCN4B PubMed Links

Gene Details

Gene
SCN4B
Gene Name
sodium voltage-gated channel beta subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142348.1 447 Missense Mutation NP_001135820.1
NM_001142349.1 447 Missense Mutation NP_001135821.1
NM_174934.3 447 Missense Mutation NP_777594.1

View Full Product Details