Product Details

SNP ID
rs150441914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56176664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTCACTGAGTTTATTCTTTTGG[A/G]ATTGACAGATCATGCTGAACTAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5J2 PubMed Links

Gene Details

Gene
OR5J2
Gene Name
olfactory receptor family 5 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005492.1 47 Missense Mutation GAA,GGA E16G NP_001005492.1

View Full Product Details