Product Details

SNP ID
rs150912530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:613820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCAGGCGCTTGGGGAGGGTGAC[A/G]GGTACGGCTCTGCCTGGTGCGGGGA
Phenotype
MIM: 606839 MIM: 605047 MIM: 611780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDHR5 PubMed Links

Gene Details

Gene
CDHR5
Gene Name
cadherin related family member 5
There are no transcripts associated with this gene.

Gene
IRF7
Gene Name
interferon regulatory factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001572.3 1261 Missense Mutation CCG,CTG P271L NP_001563.2
NM_004029.2 1261 Missense Mutation CCG,CTG P242L NP_004020.1
NM_004031.2 1261 Missense Mutation CCG,CTG P284L NP_004022.2
XM_005252906.3 1261 Missense Mutation CCG,CTG P284L XP_005252963.1
XM_005252907.3 1261 Missense Mutation CCG,CTG P283L XP_005252964.1
XM_005252909.3 1261 Missense Mutation CCG,CTG P255L XP_005252966.1
XM_011520066.2 1261 Missense Mutation CCG,CTG P270L XP_011518368.1
XM_017017674.1 1261 UTR 5 XP_016873163.1
Gene
PHRF1
Gene Name
PHD and ring finger domains 1
There are no transcripts associated with this gene.

View Full Product Details