Product Details

SNP ID
rs140151931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:12361211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGTAACTTTATTTTCAGTGACT[A/C]CTTCACCAGCCAAGCATAGAGCCAA
Phenotype
MIM: 616598
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BORCS5 PubMed Links

Gene Details

Gene
BORCS5
Gene Name
BLOC-1 related complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520551.2 80 UTR 5 XP_011518853.1
XM_011520552.2 80 Intron XP_011518854.1
XM_011520553.2 80 Missense Mutation ACT,CCT T22P XP_011518855.1
XM_017018778.1 80 Missense Mutation ACT,CCT T22P XP_016874267.1
Gene
LOH12CR2
Gene Name
loss of heterozygosity, 12, chromosomal region 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details