Product Details

SNP ID
rs142000161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:46766704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAAAGTATTTTTCCTTGTGTTA[A/G]TGATGCTTGGAATTTGGAGGATCAT
Phenotype
MIM: 608065
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC38A4 PubMed Links

Gene Details

Gene
SLC38A4
Gene Name
solute carrier family 38 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143824.1 2040 Silent Mutation CAC,CAT H547H NP_001137296.1
NM_018018.4 2040 Silent Mutation CAC,CAT H547H NP_060488.2
XM_005268997.2 2040 Silent Mutation CAC,CAT H547H XP_005269054.1

View Full Product Details