Product Details

SNP ID
rs144481515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110539917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATAATGTCTCCTAGCTGGACAT[A/C]GAAAGACGTGCTATCAGCAGCATCC
Phenotype
MIM: 609668 MIM: 608368
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPTC7 PubMed Links

Gene Details

Gene
PPTC7
Gene Name
PTC7 protein phosphatase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139283.1 859 Missense Mutation GAT,TAT D211Y NP_644812.1
Gene
RAD9B
Gene Name
RAD9 checkpoint clamp component B
There are no transcripts associated with this gene.

View Full Product Details