Product Details

SNP ID
rs144854166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21766014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATACACTCCTTCTTCCTCAGT[C/T]ACAATGGACACAAAGCGGTGGCCCC
Phenotype
MIM: 600935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNJ8 PubMed Links

Gene Details

Gene
KCNJ8
Gene Name
potassium voltage-gated channel subfamily J member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004982.3 1299 Silent Mutation GTA,GTG V328V NP_004973.1
XM_005253358.4 1299 Silent Mutation GTA,GTG V328V XP_005253415.1
XM_017019283.1 1299 Silent Mutation GTA,GTG V328V XP_016874772.1
XM_017019284.1 1299 Silent Mutation GTA,GTG V328V XP_016874773.1

View Full Product Details