Product Details

SNP ID
rs145111102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54000208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACGATGTCGGCGACGGGGCCCA[A/T]CAGTAACTATTACGTGGACTCGCTC
Phenotype
MIM: 605560 MIM: 142970 MIM: 142971 MIM: 609687
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HOXC-AS1 PubMed Links

Gene Details

Gene
HOXC-AS1
Gene Name
HOXC cluster antisense RNA 1
There are no transcripts associated with this gene.

Gene
HOXC-AS2
Gene Name
HOXC cluster antisense RNA 2
There are no transcripts associated with this gene.

Gene
HOXC10
Gene Name
homeobox C10
There are no transcripts associated with this gene.

Gene
HOXC8
Gene Name
homeobox C8
There are no transcripts associated with this gene.

Gene
HOXC9
Gene Name
homeobox C9
There are no transcripts associated with this gene.

Gene
LOC107984512
Gene Name
uncharacterized LOC107984512
There are no transcripts associated with this gene.

Gene
MIR196A2
Gene Name
microRNA 196a-2
There are no transcripts associated with this gene.

View Full Product Details