Product Details

SNP ID
rs147130309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113393094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGAGCTTCTGGATCACGTGGCT[A/G]TAGACAGCGGCCAGGGCTGCCCCGC
Phenotype
MIM: 182128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLBD2 PubMed Links

Gene Details

Gene
PLBD2
Gene Name
phospholipase B domain containing 2
There are no transcripts associated with this gene.

Gene
SDS
Gene Name
serine dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006843.2 956 Silent Mutation TAC,TAT Y278Y NP_006834.2

View Full Product Details