Product Details
- SNP ID
-
rs148173636
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:88055595 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTACCTGTTGTTCAAGGTCTTCA[C/T]TGTATTTGTTAACTTTTTGTTCTCT
- Phenotype
-
MIM: 610142
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C12orf29
PubMed Links
Gene Details
- Gene
- C12orf29
- Gene Name
- chromosome 12 open reading frame 29
There are no transcripts associated with this gene.
- Gene
- CEP290
- Gene Name
- centrosomal protein 290
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025114.3 |
8155 |
Missense Mutation |
AAT,AGT |
N2314S |
NP_079390.3 |
XM_011538756.2 |
8155 |
Missense Mutation |
AAT,AGT |
N2604S |
XP_011537058.1 |
XM_011538757.2 |
8155 |
Missense Mutation |
AAT,AGT |
N2604S |
XP_011537059.1 |
XM_011538758.2 |
8155 |
Missense Mutation |
AAT,AGT |
N2603S |
XP_011537060.1 |
XM_011538759.2 |
8155 |
Missense Mutation |
AAT,AGT |
N2601S |
XP_011537061.1 |
XM_011538760.1 |
8155 |
Missense Mutation |
AAT,AGT |
N2563S |
XP_011537062.1 |
XM_011538761.1 |
8155 |
Missense Mutation |
AAT,AGT |
N2549S |
XP_011537063.1 |
XM_011538762.2 |
8155 |
Missense Mutation |
AAT,AGT |
N2348S |
XP_011537064.1 |
XM_011538763.2 |
8155 |
Missense Mutation |
AAT,AGT |
N2317S |
XP_011537065.1 |
XM_011538764.2 |
8155 |
Intron |
|
|
XP_011537066.1 |
XM_011538765.2 |
8155 |
Intron |
|
|
XP_011537067.1 |
XM_011538766.2 |
8155 |
Missense Mutation |
AAT,AGT |
N2091S |
XP_011537068.1 |
XM_017019980.1 |
8155 |
Missense Mutation |
AAT,AGT |
N2560S |
XP_016875469.1 |
XM_017019981.1 |
8155 |
Missense Mutation |
AAT,AGT |
N2546S |
XP_016875470.1 |
XM_017019982.1 |
8155 |
Missense Mutation |
AAT,AGT |
N2604S |
XP_016875471.1 |
XM_017019983.1 |
8155 |
Missense Mutation |
AAT,AGT |
N2310S |
XP_016875472.1 |
View Full Product Details