Product Details

SNP ID
rs148234617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10908752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTGTTAGACTGAACATAGTCA[C/T]AGTGAATTTGACCGACACTGAAAAT
Phenotype
MIM: 168730 MIM: 604792
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRH1 PubMed Links

Gene Details

Gene
PRH1
Gene Name
proline rich protein HaeIII subfamily 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291314.1 811 Intron NP_001278243.1
NM_001291315.1 811 Intron NP_001278244.1
Gene
PRH1-PRR4
Gene Name
PRH1-PRR4 readthrough
There are no transcripts associated with this gene.

Gene
TAS2R13
Gene Name
taste 2 receptor member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023920.2 811 Missense Mutation ATG,GTG M183V NP_076409.1

View Full Product Details