Product Details

SNP ID
rs149232014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49349113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACCCCCTCTGAGTCCCATTCGC[C/T]TTGCTGCCCAGGTGATAGTTGGCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC22 PubMed Links

Gene Details

Gene
DNAJC22
Gene Name
DnaJ heat shock protein family (Hsp40) member C22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304944.1 983 Missense Mutation CTT,TTT L81F NP_001291873.1
NM_024902.3 983 Missense Mutation CTT,TTT L81F NP_079178.2
XM_005269155.2 983 Missense Mutation CTT,TTT L81F XP_005269212.1
XM_005269156.2 983 Missense Mutation CTT,TTT L81F XP_005269213.1
XM_005269157.2 983 Missense Mutation CTT,TTT L81F XP_005269214.1

View Full Product Details