Product Details

SNP ID
rs149379415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:15327430 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTTGCCTCCTCTTCCACATCA[A/C]CACTCCGCTATGACCACAACCAACC
Phenotype
MIM: 600579
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTPRO PubMed Links

Gene Details

Gene
PTPRO
Gene Name
protein tyrosine phosphatase, receptor type O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002848.3 Intron NP_002839.1
NM_030667.2 Intron NP_109592.1
NM_030668.2 Intron NP_109593.1
NM_030669.2 Intron NP_109594.1
NM_030670.2 Intron NP_109595.1
NM_030671.2 Intron NP_109596.1
XM_017019725.1 Intron XP_016875214.1

View Full Product Details