Product Details

SNP ID
rs150503665
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21635554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCTCACTAGTCACAGGTCTTT[C/T]AGGTCCTTCTGGATGTCCCACAGGG
Phenotype
MIM: 150100
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LDHB PubMed Links

Gene Details

Gene
LDHB
Gene Name
lactate dehydrogenase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174097.2 1326 Silent Mutation CTA,CTG L331L NP_001167568.1
NM_001315537.1 1326 Intron NP_001302466.1
NM_002300.7 1326 Intron NP_002291.1
XM_006719074.2 1326 Intron XP_006719137.1

View Full Product Details