Product Details

SNP ID
rs141407092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20731963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTCAAAACTATGTGCAGCAATT[C/T]TTTCGGGTAAGTCCAATGGATTATT
Phenotype
MIM: 607587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEF1AKMT1 PubMed Links

Gene Details

Gene
EEF1AKMT1
Gene Name
eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318939.1 507 Missense Mutation NP_001305868.1
NM_174928.2 507 Missense Mutation NP_777588.1
XM_017020432.1 507 Missense Mutation XP_016875921.1
Gene
IL17D
Gene Name
interleukin 17D
There are no transcripts associated with this gene.

View Full Product Details