Product Details

SNP ID
rs141447437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:70107577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTGCTCAAAGCTGCCACTGCC[A/G]TCCTGTTGCAGGCAGCCTCCCCCCG
Phenotype
MIM: 603680 MIM: 605332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN8OS PubMed Links

Gene Details

Gene
ATXN8OS
Gene Name
ATXN8 opposite strand (non-protein coding)
There are no transcripts associated with this gene.

Gene
KLHL1
Gene Name
kelch like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286725.1 917 Silent Mutation NP_001273654.1
NM_020866.2 917 Silent Mutation NP_065917.1
XM_017020678.1 917 Intron XP_016876167.1
XM_017020679.1 917 Intron XP_016876168.1

View Full Product Details