Product Details

SNP ID
rs142420975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39343960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTATGGGTAGTGCTTCTGTTT[C/T]TTGCCCGAAAAGCAAGCCAGCCACG
Phenotype
MIM: 606710
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LHFP PubMed Links

Gene Details

Gene
LHFP
Gene Name
lipoma HMGIC fusion partner
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005780.2 1090 Silent Mutation AAA,AAG K193K NP_005771.1
XM_011534861.1 1090 Silent Mutation AAA,AAG K193K XP_011533163.1

View Full Product Details